ContactUsLinkComponent
Safety spectacles, 600 series, SecureFit™
Safety spectacles, 600 series, SecureFit™
Catalog # MMMASF601SGAFFIN
Supplier:  3M
undefined
Safety spectacles, 600 series, SecureFit™
Catalog # MMMASF601SGAFFIN
Supplier:  3M
Specifications
  • Type:
    Scotchgard anti-fog
  • Colour:
    Grey
  • Description:
    Lens marking: 2C-1,2 3M 1 FT
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
3M™ SecureFit™ Safety glasses 600 series are semi-rimless glasses with 3M™ pressure diffusion temple technology and fixed-length temple arms over the ears.
  • Robust semi-rimless safety glasses with dual-lenses provide optimal optical performance
  • The Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalisation
  • Temples flex and self-adjust to the size of wearer's head
  • Scotchgard™ Anti-Fog coating provides superior anti-fog and anti-scratch properties (available on selected models)
  • Polarised and green IR filters available
  • Wraparound design helps a secure fit and prevents slipping
  • Foam gasket available to help limit eye exposure to debris
For optimal optical performance and good field of vision, use the SecureFit™ Safety Glasses 600 Series. These innovative and stylish semi-rimless, grey framed safety glasses have two separate lenses for a good field of vision and wide, fixed-length temple arms over the ears for a close, comfortable fit and solid side protection. They feature Pressure Diffusion Temple Technology to help diffuse pressure over the ear and soft nose pads to enhance frame comfort across a diverse workforce. The wraparound design helps provide a secure fit and prevents slipping. These glasses combine well with ear muffs.