Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Chromatography Vial Closures
Closures for high recovery vials
Closures for high recovery vials
Closures for high recovery vials
HEWL5180-0842
:
Agilent
Closures for high recovery vials
HEWL5180-0842
:
Agilent
Cap Size:
8 mm
Septum:
PTFE/silicone/PTFE
Description:
Crimp cap
Pk:
500