Complete control systems
Supplier: STRATAGENE
Certificates
About this item
The Complete control inducible mammalian expression system is designed to provide the tightest control of expression available in mammalian cells. The system is based on a synthetic ecdysone-inducible receptor and a synthetic receptor recognition element that modulates expression of your gene of interest.
- Dose-responsive inducible mammalian expression
- Induction as high as 1700 fold
- Novel IRES site allows expression of both receptor subunits from the same CMV promoter
- Retroviral and plasmid gene delivery systems available
Caution: For Research Use Only. Not for Use in Diagnostic Procedures.
Frequently Bought Together