ContactUsLinkComponent
Linking food science to biotechnology kit
Linking food science to biotechnology kit
Catalog # EDVOS-47
Supplier:  EDVOTEK
Linking food science to biotechnology kit
Catalog # EDVOS-47
Supplier:  EDVOTEK
Specifications
  • Description:
    Linking food science to biotechnology kit
  • Pk:
    1 KIT
Specifications
About this item
Investigate how agarose gel electrophoresis unlocks the color code used by food scientists to make colorful candies.
  • NGSS-aligned with MS-PS1
  • For 10 lab groups
  • Complete in 45 minutes
  • Explore the biochemistry behind colorful candies
  • Understand the biology behind gel electrophoresis
  • Perform agarose gel electrophoresis to separate different sized dye molecules
  • Extract, identify, and size different food dyes from popular candies
Students will extract color activities from common candies and separate the dyes on agarose gel electrophoresis. A fun lab extension involves the use of candy to build a DNA model.
Ordering information: DNA electrophoresis, micropipettes: 5-50 µl (optional), white light box (Optional), and microwave or hot plate are not included.
Delivery: Kit includes instructions, Ready-to-Load QuickStrip™ dye samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), practice gel loading solution, disposable pipets, and microcentrifuge tubes.
Packaging: Room temperature storage.