ContactUsLinkComponent
Ammonium acetate solution 5M
Ammonium acetate solution 5M
Catalog # R012
Supplier:  G-Biosciences
CAS Number:  
undefined
Ammonium acetate solution 5M
Catalog # R012
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Seq:
    2440
Specifications
About this item
 :
 :77.08 g/mol
Storage Temperature:Ambient
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066