ContactUsLinkComponent
Chicken Lysozyme (from egg white)
Chicken Lysozyme (from egg white)
Supplier:  Apollo Scientific
Certificates
Chicken Lysozyme (from egg white)
Supplier:  Apollo Scientific
Certificates
Specifications
  • Source:
    Egg white
  • Species:
    Chicken
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00131557
  • Enzyme name:
    Lysozyme