Aspergillus niger Glucose oxidase
APOSBIB3068-1MU
:
- Pk:1
- Species:Aspergillus niger
- Storage conditions:Freezer
- MDL No.:MFCD00131182
- Enzyme name:Glucose oxidase
- Enzyme synonyms:GOD (AN), beta-D-Glucose: oxygen 1-oxidoreductase
- Pk Info:1 MU
Lyophilized powder. From Aspergillus niger. Specific activity: IU/mg protein>300 units. IU per 1mg powder is approximately 450 units.