Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-Insulin Mouse Antibody
Anti-Insulin Mouse Antibody
Anti-Insulin Mouse Antibody
Catalog # APOSBITP2065-1MG
Supplier:
Apollo Scientific
Expand image
Anti-Insulin Mouse Antibody
Catalog # APOSBITP2065-1MG
Supplier:
Apollo Scientific
Specifications
Antigen name:
Insulin
Host:
Mouse
Reactivity:
Human
Pk:
1 mg
Specifications
About this item
Product Details
Product Details