Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-Leptin Mouse Antibody
  APOSBITP1985-1MG
Anti-Leptin Mouse Antibody
  APOSBITP1985-1MG

 

  • Antigen name:
    Leptin
  • Antigen symbol:
    Leptin
  • Host:
    Mouse
  • Reactivity:
    Human
  • Pk:
    1 mg