Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Poly(D,L-Lactide-co-trimethylene carbonate)
Poly(D,L-Lactide-co-trimethylene carbonate) 50:50
Poly(D,L-Lactide-co-trimethylene carbonate) 50:50
Poly(D,L-Lactide-co-trimethylene carbonate) 50:50
Catalog # APOSBIBE1009-5G
Supplier:
Apollo Scientific
CAS Number:
Expand image
Poly(D,L-Lactide-co-trimethylene carbonate) 50:50
Catalog # APOSBIBE1009-5G
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pack type:
Bottle
Pk:
5 g
Seq:
3173
Specifications
About this item
Product Details
:
Product Details