Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Batroxobin
Recombinant Batroxobin
Recombinant Batroxobin
APOSBITP2117-1MG
:
Apollo Scientific
:
Recombinant Batroxobin
APOSBITP2117-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Seq:
23
Protein/peptide name:
Batroxobin