1,4,7,10-Tetraazacyclododecane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:294-90-6
:
Formula:C₈H₂₀N₄
MDL Number:MFCD00066281
Melting Pt:110…113 °C
Storage Temperature:Ambient
EINECS:425-450-9
UN:1759
ADR:8,III
:172.27 g/mol
:GHS05
:GHS07
:GHS09
Frequently Bought Together