Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Propylbenzoic acid
4-Propylbenzoic acid 98%
4-Propylbenzoic acid 98%
APOSOR21930-1G
:
Apollo Scientific
:
4-Propylbenzoic acid 98%
APOSOR21930-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
164.2 g/mol
:
142…144 °C
:
GHS07
:
Ambient
:
2438-05-3
:
C₁₀H₁₂O₂
:
MFCD00013996