Order Entry
Slovenia
ContactUsLinkComponent
Holmium 99%, chips
Holmium 99%, chips
Catalog # APOSIN1963-5G
Supplier:  Apollo Scientific
CAS Number:  
Holmium 99%, chips
Catalog # APOSIN1963-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

 :
 :164.93 g/mol
Storage Temperature:Ambient
CAS Number:7440-60-0
Formula:Ho
MDL Number:MFCD00011049
Boiling Pt:2600 °C
Melting Pt:1474 °C
Density:8.8 g/cm³ (25 °C)