Orders LinkContactUsLinkComponent
Easy Trap Duster Sheets
Easy Trap Duster Sheets
Supplier:   3M
Easy Trap Duster Sheets
Supplier:  3M
Certificates
Easy Trap Duster Sheets
Easy Trap Duster Sheets
4 Options Available Below
Product Details & Documents
3M™ Easy Trap Duster system traps and holds six times more dust, dirt, sand, and hair than traditional mops and other disposable dusting systems.
  • Two-sided disposable sheets
  • Low drag reduces worker fatigue
  • Works in wet environments
  • Works on most horizontal surfaces
  • Reduce the risk of spreading germs and bacteria
Clean floors and surfaces more effectively and reliably with 3M™ Duster Sheets. These dusting sheets are made from a unique floor dusting material that can capture and hold dust and dirt, as well as hard-to-capture debris like hair and glitter. They can be used wet or dry and both sides of the sheet can be used for maximum pick up. They also help to reduce the risk of spreading germs and bacteria.
The duster sheets contain a minimum of 60% recycled content and come in perforated rolls which allow users to customise the sheet size, reducing waste. They can be used with most common cleaning tools and adapted to fit almost any flat mop system.
Product Family Options
Product Information
  • Colour
    L×W
    Availability
    Price
  • MMMAET7448
    Easy trap duster sheets
    Easy trap duster sheets
    White
    152×203 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 106,00 €
    106,00 €
    1 * 1 Roll
       106,00 €
  • MMMAET7449
    Easy trap duster sheets
    Easy trap duster sheets
    Green
    152×127 mm
    Log in for availability
    Log in for availability
    1 * 2 Roll
    1 * 2 Roll - 91,90 €
    91,90 €
    1 * 2 Roll
       91,90 €
  • MMMAET7447
    Easy trap duster sheets
    Easy trap duster sheets
    White
    152×127 mm
    Log in for availability
    Log in for availability
    1 * 8 Roll
    1 * 8 Roll - 182,00 €
    182,00 €
    1 * 8 Roll
       182,00 €
  • MMMAET7446
    Easy trap duster sheets
    Easy trap duster sheets
    White
    152×203 mm
    Log in for availability
    Log in for availability
    1 * 8 Roll
    1 * 8 Roll - 208,00 €
    208,00 €
    1 * 8 Roll
       208,00 €