ContactUsLinkComponent
PIG® HazMat Spill kit in a clip-close bag
PIG® HazMat Spill kit in a clip-close bag
Catalog # NEWPKITE391
Supplier:  NEW PIG
undefined
PIG® HazMat Spill kit in a clip-close bag
Catalog # NEWPKITE391
Supplier:  NEW PIG
Specifications
  • Description:
    Spill kit, includes 12 mat pads, 1 sock and 1 disposal bag
  • W×D×H:
    100×510×380 mm
  • Absorbency:
    13 L
  • Pk:
    1 KIT
Specifications
About this item
PIG absorbents in low cost, easy to close, hang and carry bag.
  • PIG Sock to stop spreading spills; PIG Mat absorbs quickly
  • PIG HazMat absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Temporary disposal bag helps make clean up easier
  • Durable see-through bag allows you to quickly and easily inspect spill kit supplies
  • Heavy gauge polythene bag helps protect absorbents from moisture and damage during storage to ensure maximum product performance in use
  • Convenient clip close handle makes kit easy to carry to spill site for rapid response during emergencies
The compact size makes the kit easy and convenient to store whether it’s behind the seat of a vehicle or fork truck, or hanging in spill prone areas.
Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.