ContactUsLinkComponent
Cleanroom sleeve protectors, Tyvek® IsoClean®, model IC501B 00, Non sterile
  DUPOIC501BWH00
 :  DuPont
undefined
Cleanroom sleeve protectors, Tyvek® IsoClean®, model IC501B 00, Non sterile
  DUPOIC501BWH00
 :  DuPont
  • Clothing Size:
    One size
  • Garment application:
    Cleanroom,
    Laboratory
  • Gender:
    Unisex
  • Colour:
    White
  • Pk:
    100
  • Fabric:
    Tyvek®
  • Seam type:
    Bound
Tyvek® IsoClean® sleeve model IC501B. Not clean-processed and not sterilised.
  • Bound internal seams
  • Tunnelled elastication at wrist and bicep
  • Partial body chemical protective clothing
  • Barrier to infective agents
Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms.
 : Certified according to Regulation (EU) 2016/425; Partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents).
 : Packed in cardboard box with 100 per box, bulk packed. 2 polyethylene liners.