Cleanroom boot covers, Tyvek® IsoClean®, model IC451S, non sterile
: DuPont
DuPont™ Tyvek® IsoClean® slip-retardant shoe cover model IC451S.
- Serged internal seams
- Gripper™ sole
- Not clean-processed and not sterilized
Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms.
: Certified according to Regulation (EU) 2016/425; Partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents).
: Cardboard box, 100 per box, bulk packed. 2 polyethylene liners.
- Catalog No:
- DUPOIC451SWH00/L
- DUPOIC451SWH00/M
- Fabric:
- Tyvek®
- Tyvek®
- Gender:
- Unisex
- Unisex
- Seam type:
- Serged
- Serged
- Garment application:
- Cleanroom
- Cleanroom
- Tensile strength:
- >30 N
- >30 N
Frequently Bought Together