Overalls, with hood,Tychem® 2000 C Plus
: DuPont
DuPont™ Tychem® 2000 C Plus, model CHZ5 is made from a lightweight and durable fabric (<450g per garment), Tychem® 2000 garments utilise the strength of Tyvek® and a polymeric barrier coating to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).
- Hooded coverall
- Double cuffs, thumb loops
- Elastication at wrists, ankles, face and waist
- Stitched and over-taped seams with barrier tape for protection and strength
- Front closure with double zipper, double flap, outer flap with adhesive tape
Tychem® 2000 is used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing.
: Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
Certified according to Regulation (EU) 2016/425
Antistatic treatment (EN 1149-5) - on inside
EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
Certified according to Regulation (EU) 2016/425
Antistatic treatment (EN 1149-5) - on inside
EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
Frequently Bought Together