Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amyloid Stain, Congo Red Kit
Amyloid Stain, Congo Red Kit, Sigma-Aldrich®
Amyloid Stain, Congo Red Kit, Sigma-Aldrich®
Amyloid Stain, Congo Red Kit, Sigma-Aldrich®
HT60-1KT
:
SIGMA ALDRICH MICROSCOPY
:
Amyloid Stain, Congo Red Kit, Sigma-Aldrich®
HT60-1KT
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
1 KIT
Seq:
0075
:
:
M-Clarity™ quality level = MQ500 (
more info.
)
:
2924
:
GHS02
:
GHS07
:
GHS08
:
Refrigerator
:
1