ContactUsLinkComponent
Wipe rolls, WYPALL* L20 clean and maintenance
Wipe rolls, WYPALL* L20 clean and maintenance
Catalog # KIMB72773
Supplier:  KIMBERLY CLARK
Wipe rolls, WYPALL* L20 clean and maintenance
Catalog # KIMB72773
Supplier:  KIMBERLY CLARK
Specifications
  • Packed:
    400 sheets/roll
  • Colour:
    Blue
  • Description:
    Centrefeed roll, 2-ply
  • L×W:
    380×185 mm
  • Environmentally Preferable:
  • Pk:
    6 Roll
Specifications
About this item
WypAll® wiping paper products are designed for the specific environments where they will be used, to deliver on your wiping task, every time.
  • Portable and durable
  • Embossing technology delivers a great balance of performance and value
These rolls are compatible with the quick-loading, versatile and easy-clean Aquarius™ centrefeed dispensing system (KIMB7017).