ContactUsLinkComponent
Cleanroom face masks, Kimtech™ M5
  111-0383
Cleanroom face masks, Kimtech™ M5
  111-0383
  • Type:
    With earloops
  • Material:
    Polyester/cellulose
  • Colour:
    Blue
  • Pk:
    500
Kimtech™ Pure M5 Pleat-style face masks with ear loops are a smart piece of personal protective equipment (PPE).
  • Three-layer design of polyester and cellulose, with a soft inner layer
  • The pleat-style masks have ear loops and a fully enclosed bendable nosepiece
  • Ultra sonically bonded ear loops are durable
They provide good breathability for comfort. These double bagged face masks are popular for a variety of environments, including those for biotech, medical devices, electronics and telecommunications.