ContactUsLinkComponent
Wipes, KIMTECH PREP® SCOTTPURE™
Wipes, KIMTECH PREP® SCOTTPURE™
Catalog # KIMB61930
Supplier:  KIMBERLY CLARK
undefined
Wipes, KIMTECH PREP® SCOTTPURE™
Catalog # KIMB61930
Supplier:  KIMBERLY CLARK
Specifications
  • Packed:
    Center-Pull Rolls (225 sheets per roll)
  • Colour:
    White
  • Description:
    SCOTTPURE™ wipes
  • Pk:
    6 Roll
Specifications
About this item
KIMTECH PREP® SCOTTPURE™ Wipers are designed for wiping tasks where precision and reliability are essential.
Low-lint rayon/polyester spunlace material for cloth-like softness. Engineered for excellent absorbency and low-lint wiping. Solvent resistant. No binders or surfactants. Applications include aerospace, clean manufacturing areas, DVD manufacturing, and printing.
Ordering information: Supplied without dispenser.