ContactUsLinkComponent
Universal thread adapter, for barrel and canister, V2A
Universal thread adapter, for barrel and canister, V2A
Catalog # 331-0381
Supplier:  BURKLE
undefined
Universal thread adapter, for barrel and canister, V2A
Catalog # 331-0381
Supplier:  BURKLE
Specifications
  • Hole Ø:
    48 - 75 mm
  • Description:
    Universal thread adapter, V2A
  • Pk:
    1
Specifications
About this item
Universal thread adapter for nearly all drums and other containers with opening of 48 to 75 mm in diameter, regardless of the respective thread.
The thread adapter can be mounted quickly and easily with clamping screws and can be used for different container openings. With G2" connector thread for drum screw connections and pumps with immersion tube diameter up to 32 mm.