Chromate test strips, Dosatest
Catalog # 85695.601
Supplier: VWR Chemicals
CAS Number:
Specifications
- Chemical method:Carbazide
- Pk:100
- Description:Chromate test strips
- Range:0 • 3 • 10 • 30 • 100 mg/L CrO₄²⁻
Specifications
About this item
Semi-quantitative test strips for the determination of chromate.
- Ideal for rapid and easy analysis of chromate directly on-site
- No maintenance or calibration is required
Colour change: White to violet.
Delivery: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box