Order Entry
Slovenia
ContactUsLinkComponent
Hydroxynaphthol blue, AnalaR NORMAPUR® indicator for metal titration
Irritant
Hydroxynaphthol blue, AnalaR NORMAPUR® indicator for metal titration
Catalog # 88254.130
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Hydroxynaphthol blue, AnalaR NORMAPUR® indicator for metal titration
Catalog # 88254.130
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Black powder
  • Suitability as metal indicator:
    Passes test
  • Loss on drying (105°C):
    ≤ 5 %
  • Absorption max. (pH 12.0):
    644 - 648 nm

Specification Test Results

About this item

 :
 :620.47 g/mol
EINECS:264-197-7
 :GHS07
Storage Temperature:Ambient
CAS Number:63451-35-4
Formula:C₂₀H₁₁N₂Na₃O₁₁S₃
MDL Number:MFCD00036393