Micro-volume spectrophotometers, NanoVue™ Plus
Catalog # 634-0955
Supplier: BIOCHROM
Specifications
- Detector:Charged-couple device (CCD) array
- Light source:Pulsed xenon
- Memory:90 values
- Description:Wireless spectrophotometer
- Seq:02
- W×D×H:260×390×100 mm
- Wavelength range:200 - 1100 nm
- Wavelength accuracy:±2 nm
- Absorbance accuracy:±0,01 A @ 259 nm
- Display:Backlit graphical LCD
- Interfaces:USB - Optional Bluetooth
- Spectral bandwidth:5 nm
- Weight:4,5 kg
- Pk:1
Specifications
About this item
NanoVue Plus microvolume spectrophotometers are designed for the measurement of nucleic acid and protein samples with low sample volumes.
- Hydrophobic sample plate facilitates outstanding sample recovery and reduced carryover
- Eliminates the need for cuvettes, capillaries or other sample devices
- Pre-defined methods for nucleic acid quantitation: Concentration, purity and theoretical Tm
- Pre-defined methods for protein quantitation: Direct UV, Bradford, BCA, Biuret and Lowry
- Automatic self-calibration on start up using optional path length recalibration kit
Samples of 0,5 to 2 µl can be pipetted onto the gold-coloured sample plate for measurement and then recovered using a pipette.
Delivery: Supplied with software for Print Via Computer (PVC) and for CyDye™ incorporation.