High visibility bomber jacket, RT32
About this item
This jacket is made of 300D Oxford weave polyester with a stain resistant finish and PU coating. Completely padded and lined, the jacket offers great freedom of movement.
- Waterproof with taped seams preventing water penetration
- Reflective tape for increased visibility
- Fully lined and padded to trap the heat and increase warmth
- Four pockets for ample storage
- Concealed phone pocket
- Internal chest pocket
Weight: 190 g/m²
Two-way zip for quick and easy access with storm flap in front to protect against the elements and has a concealed detachable hood.
Certifications: EN ISO 20471 Class 3, EN 343 Class 3:1, EN 342 (0.503M2. K/W(B), X, 3, 2)
9 Options Available Below
Product Details & Documents
This jacket is made of 300D Oxford weave polyester with a stain resistant finish and PU coating. Completely padded and lined, the jacket offers great freedom of movement.
- Waterproof with taped seams preventing water penetration
- Reflective tape for increased visibility
- Fully lined and padded to trap the heat and increase warmth
- Four pockets for ample storage
- Concealed phone pocket
- Internal chest pocket
Weight: 190 g/m²
Two-way zip for quick and easy access with storm flap in front to protect against the elements and has a concealed detachable hood.
Certifications: EN ISO 20471 Class 3, EN 343 Class 3:1, EN 342 (0.503M2. K/W(B), X, 3, 2)
Specifications for Product Family
Specifications
- Catalog No:
- PRTWRT32ORRXS
- PRTWRT32ORRS
- PRTWRT32ORRM
- PRTWRT32ORRL
- PRTWRT32ORRXL
- PRTWRT32ORRXXL
- PRTWRT32ORRXXXL
- PRTWRT32ORR4XL
- PRTWRT32ORR5XL
- Model:
- RT32
- RT32
- RT32
- RT32
- RT32
- RT32
- RT32
- RT32
- RT32
- Garment application:
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- Polyester/PU
- Polyester/PU
- Polyester/PU
- Polyester/PU
- Polyester/PU
- Polyester/PU
- Polyester/PU
- Polyester/PU
- Polyester/PU
- Weight:
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
Specifications
Product Family Options
Product Information
- TypeColourClothing SizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.PRTWRT32ORRXSModelRT32Garment applicationIndustrial|OutdoorRisk areaHiVisGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabricPolyester/PUWeight190 g/m² - Specifications:
More
Specifications
Cat. No.PRTWRT32ORRSModelRT32Garment applicationIndustrial|OutdoorRisk areaHiVisGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabricPolyester/PUWeight190 g/m² - Specifications:
More
Specifications
Cat. No.PRTWRT32ORRMModelRT32Garment applicationIndustrial|OutdoorRisk areaHiVisGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabricPolyester/PUWeight190 g/m² - Specifications:
More
Specifications
Cat. No.PRTWRT32ORRLModelRT32Garment applicationIndustrial|OutdoorRisk areaHiVisGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabricPolyester/PUWeight190 g/m² - Specifications:
More
Specifications
Cat. No.PRTWRT32ORRXLModelRT32Garment applicationIndustrial|OutdoorRisk areaHiVisGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabricPolyester/PUWeight190 g/m² - Specifications:
More
Specifications
Cat. No.PRTWRT32ORRXXLModelRT32Garment applicationIndustrial|OutdoorRisk areaHiVisGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabricPolyester/PUWeight190 g/m² - Specifications:
More
Specifications
Cat. No.PRTWRT32ORRXXXLModelRT32Garment applicationIndustrial|OutdoorRisk areaHiVisGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabricPolyester/PUWeight190 g/m² - Specifications:
More
Specifications
Cat. No.PRTWRT32ORR4XLModelRT32Garment applicationIndustrial|OutdoorRisk areaHiVisGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabricPolyester/PUWeight190 g/m² - Specifications:
More
Specifications
Cat. No.PRTWRT32ORR5XLModelRT32Garment applicationIndustrial|OutdoorRisk areaHiVisGenderUnisexSleeve or leg lengthLongClosure typeZip closureFabricPolyester/PUWeight190 g/m²