ContactUsLinkComponent
Delicate surface cleaning pads, Scotch-Brite™
Delicate surface cleaning pads, Scotch-Brite™
Catalog # MMMAHP98
Supplier:  3M
undefined
Delicate surface cleaning pads, Scotch-Brite™
Catalog # MMMAHP98
Supplier:  3M
Specifications
  • Colour:
    White
  • L×W:
    224×158 mm
  • Pk:
    60
Specifications
About this item
Cleaning Pads for the cleaning and efficient soft scouring of delicate surfaces.
  • Light duty scouring pads
  • Designed for use on delicate surfaces, such as porcelain, glass, and ceramic
  • Dishwasher safe
Soft, non-woven fibers and mild abrasive gently, but thoroughly cleans most surfaces without scratching.