Avantor® Hichrom HI-624, GC Columns
Supplier: Avantor
Certificates
About this item
General purpose low to mid polarity phase for a wide range of applications, including (for example): Residual solvents and volatile pollutants.
- Phase composition: Cross-bonded 6% Cyanopropyl phenyl, 94% Methyl Polysiloxane
- Long lifetime
Max. temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI08-10-020-10
- HI08-10-020-15
- HI08-10-010-15
- HI08-32-050-50
- HI08-10-040-15
- HI08-25-140-30G5
- HI08-32-100-10
- HI08-25-140-60G5
- HI08-25-025-30G5
- HI08-25-025-10
- HI08-32-100-105
- HI08-10-040-10
- HI08-25-025-60G5
- HI08-53-300-30G5
- HI08-25-025-105
- HI08-15-015-10
- HI08-32-150-10
- HI08-32-180-30G5
- HI08-53-100-30G5
- HI08-25-050-10
- HI08-53-150-10
- HI08-53-100-75
- HI08-25-025-60
- HI08-32-150-105
- HI08-53-150-30
- HI08-25-050-15
- HI08-53-150-15
- HI08-25-050-105
- HI08-53-050-60
- HI08-53-050-50
- HI08-25-010-50
- HI08-53-100-15
- HI08-53-100-30
- HIGCHI08-20-112-25
- HI08-32-025-105
- HI08-32-025-15
- HI08-10-010-10
- HI08-32-025-50
- HI08-32-050-105
- HI08-05-005-2.5
- HI08-20-112-40
- HI08-53-025-50
- HI08-05-005-5
- HI08-32-050-30
- HI08-53-050-10
- HI08-25-025-15G5
- HI08-20-112-50
- HI08-53-025-60
- HI08-53-050-15
- HI08-18-100-20
- HI08-32-300-60
- HI08-18-040-50
- HI08-18-100-40
- HI08-53-025-30
- HI08-53-025-15
- HIGCHI0853300105
- HI08-25-300-30
- HI08-18-020-20
- HI08-18-020-40
- HI08-32-010-105
- HI08-32-010-50
- HI08-32-010-30
- HI08-32-300-105
- HI08-18-040-40
- HI08-18-040-20
- HI08-32-300-30
- HI08-18-018-40
- HI08-32-150-30
- HI08-32-150-60
- HI08-32-150-50
- HI08-32-180-30
- HI08-25-050-60
- HI08-32-180-105
- HI08-15-084-40
- HI08-25-050-50
- HI08-25-100-10
- HI08-25-100-50
- HI08-25-100-30
- HI08-32-180-75
- HI08-25-140-105
- HI08-25-100-60
- HI08-18-018-20
- HI08-25-140-50
- HI08-18-018-10
- HI08-25-140-30
- Phase name:
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- HI-624
- Phase composition:
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- 6% Cyanopropylphenyl, 94% Methyl Polysiloxane
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- USP code:
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43
- G43