Order Entry
Slovenia
ContactUsLinkComponent
 
Organic reference standard, Isophorone 5,000 µg/ml in methanol
  PROOU-EPA-1132
 :  
Organic reference standard, Isophorone 5,000 µg/ml in methanol
  PROOU-EPA-1132
 :  

 

  • Pk:
    1
  • CAS No.:
    78-59-1
  • Description:
    Isophorone 5,000 µg/ml in methanol
  • Formula:
    C₉H₁₄O
  • Description:
    Isophorone