Order Entry
Slovenia
ContactUsLinkComponent
Organic reference standard, Acetophenone 5000 µg/ml in methanol
Organic reference standard, Acetophenone 5000 µg/ml in methanol
Catalog # PROOU-EPA-1066
CAS Number:  
Organic reference standard, Acetophenone 5000 µg/ml in methanol
Catalog # PROOU-EPA-1066
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    98-86-2
  • Description:
    Acetophenone 5,000 µg/ml in methanol
  • Formula:
    C₈H₈O
  • Description:
    Acetophenone

Specifications

About this item