Organic reference standard, 4-Nitrophenol 5000 µg/ml in methanol
Catalog # PROOU-EPA-1141
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- CAS No.:100-02-7
- Description:4-Nitrophenol 5,000 µg/ml in methanol
- Formula:C₆H₅NO₃
- Description:4-Nitrophenol