ContactUsLinkComponent
Sodium chloride regeneration salt, TECHNICAL for water softening
Sodium chloride regeneration salt, TECHNICAL for water softening
Catalog # 99062.4000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium chloride regeneration salt, TECHNICAL for water softening
Catalog # 99062.4000
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bucket
  • Pk:
    4 kg
  • Seq:
    4102
Specifications
Specification Test Results
  • Identification:
    Passes test
Specification Test Results
About this item
 :
 :58.44 g/mol
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477