ContactUsLinkComponent
Microscope stands, basic
  630-1159
undefined
Microscope stands, basic
  630-1159
  • Description:
    MS12C, acrylic holder with two variable view range adapters, for tabletop use, suitable for all Dino-Lite models, except models in AD series and models with polariser
  • Seq:
    901
  • Pk:
    1
Holders and basic stands for Dino-Lite microscopes.
MS12C: The MS12C consists of a base unit with two insert adapters providing three 'hands free' fixed working distance and magnification options. Ideal for collectors, hobbyists, graphic artists a well as at home or in the classroom. The Dino-Lite digital microscopes simply slides into the base or one of the adapters. The clear acrylic base design allows easy accessibility of tools and light to the observed object. Supplied with two variable view range adapters, transparent.
Suitable for: All Dino-Lite models, except models in AD series and models with polariser.
MS23B: This desktop clamp with flexible gooseneck can be clamped to a desktop or any other surface of up to 3 cm thick. The arm has a wide reach of 35 cm and can be bent and twisted in the required position for easy and convenient inspection of all sorts of objects with the Dino-Lite digital microscope.
Suitable for: All Dino-Lite models.