Microarray molds kits, T-Sue™
About this item
T-Sue™ Microarray mold, enabling tissues from many donor blocks to be arrayed on a single slide.
- Can be reused hundreds of times without losing its flexibility
- Withstands temperatures between –100 to +250 °C
- View many different samples on the same slide
- Array mold is designed to be simple and easy to use
The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-made paraffin recipient block. No specific technical training or experience is needed.
They are ready to accept cores from a donor block, and offer a consistent core diameter. The cassette is permanently bonded to the block.T-Sue™ punch needles are used for filling recipient tissue microarray paraffin blocks. Internal stylet operates like a retractable pen. Effortless ejection of core specimens.
Tissues can be analyzed in the same conditions enhancing the efficiency of the research. The T-Sue™ Microarray molds allow users to perform TMAs faster while giving excellent results.
: Each kit includes ONE T-Sue™ array mold and FOUR punch needles with stylet allowing easy removal of tissue cores for insertion.
19 Options Available Below
Product Details & Documents
T-Sue™ Microarray mold, enabling tissues from many donor blocks to be arrayed on a single slide.
- Can be reused hundreds of times without losing its flexibility
- Withstands temperatures between –100 to +250 °C
- View many different samples on the same slide
- Array mold is designed to be simple and easy to use
The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-made paraffin recipient block. No specific technical training or experience is needed.
They are ready to accept cores from a donor block, and offer a consistent core diameter. The cassette is permanently bonded to the block.T-Sue™ punch needles are used for filling recipient tissue microarray paraffin blocks. Internal stylet operates like a retractable pen. Effortless ejection of core specimens.
Tissues can be analyzed in the same conditions enhancing the efficiency of the research. The T-Sue™ Microarray molds allow users to perform TMAs faster while giving excellent results.
: Each kit includes ONE T-Sue™ array mold and FOUR punch needles with stylet allowing easy removal of tissue cores for insertion.
Product Family Options
T-Sue™ Microarray molds
Product Information
- AvailabilityPrice
- SIMPM473-15T-Sue™ microarray mold, 15 mold cores, 4 mm core diameterT-Sue™ microarray mold, 15 mold cores, 4 mm core diameterLog in for availability1 * 1 items1 560,00 €
- SIMPM473-24T-Sue™ microarray mold, 24 mold cores, 2 mm core diameterT-Sue™ microarray mold, 24 mold cores, 2 mm core diameterLog in for availability1 * 1 items1 780,00 €
- SIMPM473-36T-Sue™ microarray mold, 36 mold cores, 3 mm core diameterT-Sue™ microarray mold, 36 mold cores, 3 mm core diameterLog in for availability1 * 1 items1 780,00 €
- SIMPM473-60T-Sue™ microarray mold, 60 mold cores, 2 mm core diameterT-Sue™ microarray mold, 60 mold cores, 2 mm core diameterLog in for availability1 * 1 items1 780,00 €
- SIMPM473-72T-Sue™ microarray mold, 72 mold cores, 1,5 mm core diameterT-Sue™ microarray mold, 72 mold cores, 1,5 mm core diameterLog in for availability1 * 1 items1 780,00 €
- SIMPM473-150T-Sue™ microarray mold, 150 mold cores, 1,5 mm core diameterT-Sue™ microarray mold, 150 mold cores, 1,5 mm core diameterLog in for availability1 * 1 items1 560,00 €
- SIMPM473-170T-Sue™ microarray mold, 170 mold cores, 1 mm core diameterT-Sue™ microarray mold, 170 mold cores, 1 mm core diameterLog in for availability1 * 1 items1 780,00 €
Precast T-Sue™ paraffin blocks
Product Information
- AvailabilityPrice
- SIMPM473-15PCPrecast T-Sue™ paraffin block, 15 mold cores, 4 mm core diameterPrecast T-Sue™ paraffin block, 15 mold cores, 4 mm core diameterLog in for availability1 * 6 items422,00 €
- SIMPM473-24PCPrecast T-Sue™ paraffin block, 24 mold cores, 2 mm core diameterPrecast T-Sue™ paraffin block, 24 mold cores, 2 mm core diameterLog in for availability1 * 6 items422,00 €
- SIMPM473-36PCPrecast T-Sue™ paraffin block, 36 mold cores, 3 mm core diameterPrecast T-Sue™ paraffin block, 36 mold cores, 3 mm core diameterLog in for availability1 * 6 items422,00 €
- SIMPM473-60PCPrecast T-Sue™ paraffin block, 60 mold cores, 2 mm core diameterPrecast T-Sue™ paraffin block, 60 mold cores, 2 mm core diameterLog in for availability1 * 6 items422,00 €
- SIMPM473-72PCPrecast T-Sue™ paraffin block, 72 mold cores, 1,5 mm core diameterPrecast T-Sue™ paraffin block, 72 mold cores, 1,5 mm core diameterLog in for availability1 * 6 items422,00 €
- SIMPM473-150PCPrecast T-Sue™ paraffin block, 150 mold cores, 1,5 mm core diameterPrecast T-Sue™ paraffin block, 150 mold cores, 1,5 mm core diameterLog in for availability1 * 6 items422,00 €
- SIMPM473-170PCPrecast T-Sue™ paraffin block, 170 mold cores, 1 mm core diameterPrecast T-Sue™ paraffin block, 170 mold cores, 1 mm core diameterLog in for availability1 * 6 items422,00 €
T-Sue™ punch needles
Product Information
- AvailabilityPrice
- SIMPM473-P10MAT-Sue™ punch needles, 1 mm diameter, magenta
- SIMPM473-P15YT-Sue™ punch needles, 1,5 mm diameter, yellow
- SIMPM473-P20RT-Sue™ punch needles, 2 mm diameter, red
- SIMPM473-P30PT-Sue™ punch needles, 3 mm diameter, pink
- SIMPM473-P40BT-Sue™ punch needles, 4 mm diameter, blue