ContactUsLinkComponent
Disposable particle filtering respirators, FFP2, P251
Disposable particle filtering respirators, FFP2, P251
Catalog # PRTWP251WHR
Supplier:  Portwest
undefined
Disposable particle filtering respirators, FFP2, P251
Catalog # PRTWP251WHR
Supplier:  Portwest
Specifications
  • Type:
    P251
  • Pk:
    20
  • With valve:
    +
  • Class:
    FFP2
Specifications
About this item
Vertical flat folded mask with exhalation valve for lighter breathing resistance. Ideal for all day use.
  • Adjustable nose clip for optimised fit
  • Ultrasound welded elastic bands
  • Flexible shape allows the mask to be folded flat
  • Individually packed for vending machines
Certifications: EN 149:2001 + A1:2009.
Packaging: Individually wrapped inside hygienic polybag.