ContactUsLinkComponent
VWR® TITRINORM®, pH Buffers, for Karl Fischer Titrations
VWR® TITRINORM®, pH Buffers, for Karl Fischer Titrations
Catalog # 85497.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® TITRINORM®, pH Buffers, for Karl Fischer Titrations
Catalog # 85497.260
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Acid buffer, 5 mmol acid/ml
Specifications
About this item
The volumetric and coulometric reagents are buffered systems, stabilised around the ideal value between 5 to 7 ensuring a rapid and stoichiometric course of reaction. Buffering systems which neutralise and stabilise the pH during the reaction are suitable for this purpose.