Zorbax CN, HPLC Columns
: Avantor
Zorbax spherical particles are manufactured from small extremely uniform colloidal silica micro beads which are agglutinated in a Du Pont patented organic polymerisation process. The organic polymer is removed by a controlled sintering process to yield mechanically stable silica particles.
- Spherical porous silica
- High efficiency
- Narrow particles and pore size distribution
- Robust with good batch to batch reproducibility
Zorbax spherical particles are manufactured from small extremely uniform colloidal silica micro beads which are agglutinated in a Du Pont patented organic polymerisation process. The organic polymer is removed by a controlled sintering process to yield mechanically stable silica particles.
- Spherical porous silica
- High efficiency
- Narrow particles and pore size distribution
- Robust with good batch to batch reproducibility
- Catalog No:
- ZCN-150AS
- ZCN-250AF
- ZCN-150A
- ZCN-250A
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 7%
- 7%
- 7%
- 7%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Product Family Options
Zorbax Medium bore columns
- PhasePore sizeParticle SizeØ int.Length
Low Manufacturing Impact Specifications
Cat. No.ZCN-150ASEnvironmentally PreferableLow Manufacturing ImpactCarbon load7%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Zorbax Analytical columns
- PhasePore sizeParticle SizeØ int.Length
Low Manufacturing Impact Specifications
Cat. No.ZCN-250AFEnvironmentally PreferableLow Manufacturing ImpactCarbon load7%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.ZCN-150AEnvironmentally PreferableLow Manufacturing ImpactCarbon load7%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.ZCN-250AEnvironmentally PreferableLow Manufacturing ImpactCarbon load7%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact