ContactUsLinkComponent
Goggles, pheos cx2 sonic
Goggles, pheos cx2 sonic
Catalog # UVEX9309.275
Supplier:  UVEX
undefined
Goggles, pheos cx2 sonic
Catalog # UVEX9309.275
Supplier:  UVEX
Specifications
  • Type:
    Anti-UV, anti-scratch, anti-fog
  • Colour:
    White/black
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
With its compact, lightweight design, the uvex pheos cx2 sonic is a welcome companion for any job.
  • Comfortable protection thanks to soft forehead and nose piece moulded directly onto the lens
  • Scratch-proof exterior, permanent anti-fog interior (uvex supravision extreme)
  • Can be easily combined with other PPE
  • Textile headband
These functional wide-vision goggles offer impressive comfort and, thanks to uvex supravision coating technology, provide a clear field of vision even in extreme climates.
Certifications: EN 166; EN 170