ContactUsLinkComponent
Aprons, embossed, LDPE
Aprons, embossed, LDPE
Catalog # 113-9324
Supplier:  VWR Collection
undefined
Aprons, embossed, LDPE
Catalog # 113-9324
Supplier:  VWR Collection
Specifications
  • Colour:
    Blue
  • Description:
    Blocked, thickness: 20 µm
  • Seq:
    03
  • Pk:
    1.000
  • W×L:
    800×1250 mm
Specifications
About this item
Aprons are made of low-density polyethylene (LDPE).
Packaging: 100 pieces per bag, 1000 pieces per carton.