ContactUsLinkComponent
Qualitative fit test respirator
Qualitative fit test respirator
Catalog # MMMAFT30
Supplier:  3M
undefined
Qualitative fit test respirator
Catalog # MMMAFT30
Supplier:  3M
Specifications
  • Description:
    Qualitative fit test apparatus, bitter
  • Pk:
    1
Specifications
About this item
Fit test kit for respirators.
This kit uses a test solution of denatonium benzoate to produce a very bitter taste. It can be used to check face-to-respirator seal on any particulate respirator or gas/vapour respirator with a particulate prefilter.
Certifications: Meets the performance criteria for fit testing respirators under the current OSHA standard for respirator protection: 29 CFR 1910.134, Appendix A.
Delivery: The kit includes hood, collar, two nebulisers, bitter tasting sensitivity solution, bitter tasting fit test solution and laminated user instructions.