Capsule filters, Polycap™ AS, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
These Polycap™ AS (aqueous solutions) capsule filters combine a glass microfiber (GMF) prefilter over a nylon membrane final filter to provide maximum filtration of difficult, particulate-laden solutions.
- First layer (GMF) acts as a prefilter to ensure longer membrane life and higher filtration efficiency
- Nylon membrane filter layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates
- PP housing is thermally fused (no glues, adhesives, or extraneous materials), reducing extractable levels
- Can be tested with bubble point, pressure decay, or forward flow methods
- Available in presterilised and non sterile (autoclavable) versions
Polycap™ AS are capsule filters specifically designed for high-volume filtration of aqueous sample solutions in a compact size.
Certifications: Manufactured according to ISO9001:2008 QMS standards
Specifications
- Catalog No:
- 516-0966
- 516-4445
- 515-0108
- 516-0443
- 516-0957
- 516-4436
- 516-4442
- 516-0965
- Type:
- Polycap™ AS 75, nylon, with glass fibre prefilter, non sterile
- Polycap™ AS 36, nylon, with glass fibre prefilter, sterile
- Polycap™ AS 36, nylon, with glass fibre prefilter, sterile
- Polycap™ AS 36, nylon, with glass fibre prefilter, sterile
- Polycap™ AS 36, nylon, with glass fibre prefilter, non sterile
- Polycap™ AS 36, nylon, with glass fibre prefilter, sterile
- Polycap™ AS 36, nylon, with glass fibre prefilter and filling bell, sterile
- Polycap™ AS 75, nylon, with glass fibre prefilter, non sterile
- Filtration area:
- 820 cm²
- 400 cm²
- 400 cm²
- 400 cm²
- 400 cm²
- 400 cm²
- 400 cm²
- 820 cm²
- Max. pressure:
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar