Test strips, qualitative, indanthrene yellow
Catalog # MANA90751
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Pk:200
- Description:Indanthrene yellow paper
Specifications
About this item
The test paper allows the easy determination of hydro sulphite (Sodium dithionite). It is used to determine the endpoint in the conversion of vat dyes to the leuco form. Because of the special purpose, indanthrene yellow paper is also called hydrosulphite paper.
- Colour change: Yellow to blue
Packaging: Pack of 200 strips 20×70 mm