ContactUsLinkComponent
Amino acid analysis columns, InfinityLab AdvanceBio
Amino acid analysis columns, InfinityLab AdvanceBio
Supplier:  Agilent
Certificates
Amino acid analysis columns, InfinityLab AdvanceBio
Supplier:  Agilent
Specifications
  • Catalog No:
  • HEWL823750-946
  • HEWL820750-931
  • HEWL695975-322
  • HEWL655950-802
  • Endcapped:
  • Double
  • Double
  • Double
  • Double
  • Max. pressure:
  • 600 Bar
  • 600 Bar
  • 600 Bar
  • 600 Bar
  • pH range:
  • 3,0 – 11,0
  • 3,0 – 11,0
  • 3,0 – 11,0
  • 3,0 – 11,0
  • Temperature limit:
  • 60 ˚C
  • 60 ˚C
  • 60 ˚C
  • 60 ˚C