Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vials
VWR® La-Pha-Pack™, Screw neck vials
VWR® La-Pha-Pack™, Screw neck vials
VWR® La-Pha-Pack™, Screw neck vials
Catalog # LAPH18091009
Supplier:
LA PHA PACK
Expand image
VWR® La-Pha-Pack™, Screw neck vials
Catalog # LAPH18091009
Supplier:
LA PHA PACK
Specifications
Cap included:
No
Ø×H:
22×37,2 mm
Capacity:
7,00 ml
Colour:
Clear
Material:
Glass
Pk:
100
Specifications
About this item
Product Details
Product Details