Orders LinkContactUsLinkComponent
Overhead stirrer tools
Overhead stirrer tools
Supplier:   witeg Labortechnik
Overhead stirrer tools
Supplier:  witeg Labortechnik
Certificates
Overhead stirrer tools
Overhead stirrer tools
3 Options Available Below
Product Details & Documents
These high-speed overhead stirrer tools are designed for mixing applications in the laboratory specialised for low- to medium-viscosity materials.
  • Jog-Shuttle controller with touch-button
  • Maximal viscosity 10000/50000 mPas
  • Chrome-plated stainless-steel chuck to prevent corrosion
  • Impeller height adjustable
  • Six light sources and LCD display
These units provide steady and constant stirring speed and designed with powerful DC motor for various stirring applications such as direct-driven or worm-geared.
The direct driven motor ensures high-speed at all recommended viscosities. Features with anti-trembling system that is the impeller is fixed at two points within the body and chuck.
These units are features with timer (99 h 59 min) with timer-end signal, locking mode, error status display, overload and overheat protection and operation LEDs.
Ordering information: Available as set with impeller, plate stand and clamp, many different impellers and plate stands available.
Product Family Options
Product Information
  • Shaft Ø
    Height
    Width
    Length
    Max. revolutions
    Availability
    Price
  • WITGDH.WOS01016
    Overhead stirrer, HS-30 D set, 10 L, 230 V
    Overhead stirrer, HS-30 D set, 10 L, 230 V
    8 mm
    180 mm
    90 mm
    145 mm
    200 - 3000 min-1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 701,00 €
    701,00 €
    1 * 1 items
       701,00 €
  • WITGDH.WOS01025
    Overhead stirrer, HS-100 D unit, 20 L, 230 V
    Overhead stirrer, HS-100 D unit, 20 L, 230 V
    8 mm
    180 mm
    90 mm
    145 mm
    200 - 3000 min-1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 591,00 €
    591,00 €
    1 * 1 items
       591,00 €
  • WITGDH.WOS01026
    Overhead stirrer, HS-100 D set, 20 L, 230 V
    Overhead stirrer, HS-100 D set, 20 L, 230 V
    8 mm
    180 mm
    90 mm
    145 mm
    200 - 3000 min-1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 767,00 €
    767,00 €
    1 * 1 items
       767,00 €