Orders LinkContactUsLinkComponent
Compact automatic burettes, Schellbach, BLAUBRAND®
Compact automatic burettes, Schellbach, BLAUBRAND®
50 ml, 0,10 ml
Catalog # BRND23872
Supplier:   Brand
Compact automatic burettes, Schellbach, BLAUBRAND®
Catalog # BRND23872
Supplier:  Brand
Compact automatic burettes, Schellbach, BLAUBRAND®
Compact automatic burettes, Schellbach, BLAUBRAND®
50 ml, 0,10 ml
Price
168,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3, class AS, DE-M marking.
Delivery information: Burettes supplied with 1000 ml PE bottle.
Specifications
  • Capacity:
    50 ml
  • Seq:
    03
  • Division:
    0,10 ml