Orders LinkContactUsLinkComponent
HCCA matrix
HCCA matrix
HCCA (α-Cyano-4-hydroxycinnamic acid) matrix, 10 vials, for approx 200 sample positions, 10
Catalog # BRUK8255344
Supplier:   BRUKER DALTRONIK
HCCA matrix
Catalog # BRUK8255344
Supplier:  BRUKER DALTRONIK
CAS Number:  
HCCA matrix
HCCA (α-Cyano-4-hydroxycinnamic acid) matrix, 10 vials, for approx 200 sample positions, 10
CAS Number:  
Price
983,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Pre-portioned HCCA matrix provides easy and convenient preparation of HCCA matrix solutions.
  • Stable and easy to handle for sample preparation
HCCA has been developed especially for Bruker MALDI instruments and purified to ensure accurate results and maintain system cleanliness. This reduces frequency of preventive maintenance cleans required.
Documents
Specifications
Specifications
  • Pk:
    10
  • Description:
    HCCA (α-Cyano-4-hydroxycinnamic acid) matrix, 10 vials, for approx 200 sample positions
Specifications