- Packed:100/canister, 10 canisters/case
- Description:SAT wipes
- L×W:150×230 mm
- Pk:1.000
Contec HydroPure is a blend of 6% hydrogen peroxide and purified water. Contec HydroPure is BPR authorised at EU level and efficacious against bacteria, fungi, moulds, and yeasts.
- Impregnated wipes at point-of-use provide convenient and controlled application of disinfectant
- Hydrogen Peroxide breaks down to water & oxygen leaving no residue
- 100 polyester / cellulose wipes in a canister
- Filtered to 0.2 microns in a Grade B cleanroom
- Ideal for a Grade A / B areas
- Not classed as corrosive
- Double bagged in linear tear packaging
Contec HydroPure leaves little-to-no residue making it ideal for use in critical areas. Contec HydroPure is provided as a point-of-use SATWIPES system. Each double bagged pack contains a canister of 100 x 54gsm Amplitude Theta polyester/ cellulose dry wipes and a 500ml capped bottle of Contec HydroPure.
Simply add 500 ml of HydroPure to the dry roll, 5 mins before use, and use the impregnated wipes within 7 days.
Contec HydroPure is 0.2 micron filtered, and filled in a Grade A environment. The sterile product is filtered into pre-irradiated components. The dry wipe canister is irradiated at no less than 25 kGy
*The product(s) on this page are outside the scope of the EU Directive 2019/904